missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human PRG-1/LPPR4 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2874
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PRG-1/LPPR4 Monoclonal specifically detects PRG-1/LPPR4 in Human samples. It is validated for Western Blot.
Tekniske data
| PRG-1/LPPR4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| AAO85400 | |
| LPPR4 | |
| E. coli-derived recombinant human PRG-1/LPPR4 Ser285-Asp715 Accession # AAO85400 | |
| 100 μg | |
| Primary | |
| Detects PRG-1/LPPR4 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 368701 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| LPPR4, LPPR4 lipid phosphate phosphatase-related protein type 4, LPR4, PHP1, PRG1, PRG-1, RP4-788L13.1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9890 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion