Learn More
Abnova™ Human POLRMT Partial ORF (NP_005026.2, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005442-Q02.25ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a mitochondrial DNA-directed RNA polymerase. The gene product is responsible for mitochondrial gene expression as well as for providing RNA primers for initiation of replication of the mitochondrial genome. Although this polypeptide has the same function as the three nuclear DNA-directed RNA polymerases, it is more closely related to RNA polymerases of phage and mitochondrial polymerases of lower eukaryotes. [provided by RefSeq]
Sequence: MSALCWGRGAAGLKRALRPCGRPGLPGKEGTAGGVCGPRRSSSASPQEQDQDRRKDWGHVELLEVLQARVRQLQAESVSEVVVNRVDVARLPECGSGDGSSpecifications
NP_005026.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSALCWGRGAAGLKRALRPCGRPGLPGKEGTAGGVCGPRRSSSASPQEQDQDRRKDWGHVELLEVLQARVRQLQAESVSEVVVNRVDVARLPECGSGDGS | |
RUO | |
POLRMT | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5442 | |
POLRMT (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APOLMT/MTRNAP/MTRPOL/h-mtRPOL | |
POLRMT | |
Recombinant | |
wheat germ expression system |