missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human Plasminogen Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1939
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Plasminogen Monoclonal specifically detects Plasminogen in Human samples. It is validated for Immunoprecipitation, Neutralization.
Tekniske data
| Plasminogen | |
| Monoclonal | |
| LYOPH | |
| Immunoprecipitation 25 ug/mL, Neutralization 1 ug/mL | |
| DKFZp779M0222, EC 3.4.21, EC 3.4.21.7, plasmin, plasminogen, Plg | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5340 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunoprecipitation, Neutralization | |
| 270409 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PLG | |
| Human plasma-derived Plasminogen Glu20-Asn810 | |
| 100 μg | |
| Primary | |
| Detects human PLG, human Plasmin, and the catalytic domain in direct ELISAs and Western blots. | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion