Learn More
Abnova™ Human PIWIL1 Partial ORF (NP_004755.2, 431 a.a. - 541 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009271-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the PIWI subfamily of Argonaute proteins, evolutionarily conserved proteins containing both PAZ and Piwi motifs that play important roles in stem cell self-renewal, RNA silencing, and translational regulation in diverse organisms. The encoded protein may play a role as an intrinsic regulator of the self-renewal capacity of germline and hematopoietic stem cells. [provided by RefSeq]
Sequence: NDNVQRELRDWGLSFDSNLLSFSGRILQTEKIHQGGKTFDYNPQFADWSKETRGAPLISVKPLDNWLLIYTRRNYEAANSLIQNLFKVTPAMGMQMRKAIMIEVDDRTEAYSpecifications
NP_004755.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.95kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NDNVQRELRDWGLSFDSNLLSFSGRILQTEKIHQGGKTFDYNPQFADWSKETRGAPLISVKPLDNWLLIYTRRNYEAANSLIQNLFKVTPAMGMQMRKAIMIEVDDRTEAY | |
RUO | |
PIWIL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9271 | |
PIWIL1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HIWI/MIWI/PIWI | |
PIWIL1 | |
Recombinant | |
wheat germ expression system |