missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PIST Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP0294

Additional Details : Weight : 0.01000kg

Product Code. 15266509

  • 1004.00 DKK / 50µg
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PIST Synthetic Peptide, PEP-0294, from Invitrogen.

This peptide corresponds to 16 amino acids near the carboxy terminus of human PIST.PEP-0294 can be used as a blocking peptide with polyclonal antibody PA5-20179.
Specifications

Specifications

57120
purified
Human PIST
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q9HD26
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
GOPC
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers