Learn More
Abnova™ Human PIP5K2B Full-length ORF (AAH27459, 1 a.a. - 281 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008396-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene catalyzes the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-5,4-bisphosphate. This gene is a member of the phosphatidylinositol-5-phosphate 4-kinase family. The encoded protein sequence does not show similarity to other kinases, but the protein does exhibit kinase activity. Additionally, the encoded protein interacts with p55 TNF receptor. [provided by RefSeq]
Sequence: MSSNCTSTTAVAVAPLSASKTKTKKKHFVCQKVKLFRASEPILSVLMWGVNHTINELSNVPVPVMLMPDDFKAYSKIKVDNHLFNKENLPSRFKFKEYCPMVFRNLRERFGIDDQDYQNSVTRSAPINSDSQGRCGTRFLTTYDRRFVIKTVSSEDVAEMHNILKKYHQFIVECHGNTLLPQFLGMYRLTVDGVETYMVVTRNVFSHRLTVHRKYDLKGSTVAREASDKEKAKDLPTFKDNDFLNEGQKLHVGEESKKNFLEKLKRDVEEILVLSPGRRIASpecifications
AAH27459 | |
Liquid | |
8396 | |
PIP5K2B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSSNCTSTTAVAVAPLSASKTKTKKKHFVCQKVKLFRASEPILSVLMWGVNHTINELSNVPVPVMLMPDDFKAYSKIKVDNHLFNKENLPSRFKFKEYCPMVFRNLRERFGIDDQDYQNSVTRSAPINSDSQGRCGTRFLTTYDRRFVIKTVSSEDVAEMHNILKKYHQFIVECHGNTLLPQFLGMYRLTVDGVETYMVVTRNVFSHRLTVHRKYDLKGSTVAREASDKEKAKDLPTFKDNDFLNEGQKLHVGEESKKNFLEKLKRDVEEILVLSPGRRIA | |
RUO | |
PIP4K2B | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
56.65kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PI5P4KB/PIP5K2B/PIP5KIIB/PIP5KIIbeta | |
PIP4K2B | |
Yes | |
wheat germ expression system |