Få mere at vide
Human Phospho-p27/Kip1 (T157) Antibody, R&D Systems™
Beskrivelse
p27/Kip1 Polyclonal specifically detects p27/Kip1 in Human samples. It is validated for Western Blot.
Tekniske data
Tekniske data
| Antigen | p27/Kip1 (p thr157) |
| Applikationer | Western Blot |
| Klassifikation | Polyclonal |
| Koncentration | LYOPH |
| Konjugeret | Unconjugated |
| Fortynding | Western Blot 0.5 ug/mL |
| Gene Alias | CDKN1B, CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, Kip1, KIP1P27KIP1, MEN1B, MEN4, p27Kip1 |
| Gensymboler | CDKN1B |
| Værtsarter | Rabbit |
| Immunogen | Phosphopeptide containing human p27/Kip1 T157 site |
| Vis mere |