missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PGF Full-length ORF (NP_002623.2, 19 a.a. - 170 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005228-P02L

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132695

  • 4040.00 DKK / 25µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: LPAVPPQQWALSAGNGSSEVEVVPFQEVWGRSYCRALERLVDVVSEYPSEVEHMFSPSCVSLLRCTGCCGDENLHCVPVETANVTMQLLKIRSGDRPSYVELTFSQHVRCECRPLREKMKPERRRPKGRGKRRREKQRPTDCHLCGDAVPRR
Specifications

Specifications

NP_002623.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
D12S1900/PGFL/PLGF/PlGF-2/SHGC-10760
PGF
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5228
PGF (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue
LPAVPPQQWALSAGNGSSEVEVVPFQEVWGRSYCRALERLVDVVSEYPSEVEHMFSPSCVSLLRCTGCCGDENLHCVPVETANVTMQLLKIRSGDRPSYVELTFSQHVRCECRPLREKMKPERRRPKGRGKRRREKQRPTDCHLCGDAVPRR
RUO
PGF
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers