Learn More
Abcam Human PDGFRA knockout SH-SY5Y cell line
Product Code. 32157602 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32157602 1,000,000 Cells Each
32142603 2 x 1,000,000 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32157602 Supplier Abcam Supplier No. ab2753351000000Cells_
 Restricted Item
Forventet forsendelse: 07-12-2026
PDGFRA KO cell line available to order. KO validated by Immunocytochemistry, Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 67 bp deletion in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial....
PDGFRA KO cell line available to order. KO validated by Immunocytochemistry, Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 67 bp deletion in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1,000,000 Cells
Regulatory Status RUO