Learn More
Abnova™ Human PDCL Full-length ORF (AAH17227, 1 a.a. - 301 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005082-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Phosducin-like protein is a putative modulator of heterotrimeric G proteins. The protein shares extensive amino acid sequence homology with phosducin, a phosphoprotein expressed in retina and pineal gland. Both phosducin-like protein and phosphoducin have been shown to regulate G-protein signaling by binding to the beta-gamma subunits of G proteins. [provided by RefSeq]
Sequence: MTTLDDKLLGEKLQYYYSSSEDEDSDHEDKDRGRCAPASSSVPAEAELAGEGISVNTGPKGVINDWRRFKQLETEQREEQCREMERLIKKLSMTCRSHLDEEEEQQKQKDLQEKISGKMTLKEFAIMNEDQDDEEFLQQYRKQRMEEMRQQLHKGPQFKQVFEISSGEGFLDMIDKEQKSIVIMVHIYEDGIPGTEAMNGCMICLAAEYPAVKFCKVKSSVIGASSQFTRNALPALLIYKGGELIGNFVRVTDQLGDDFFAVDLEAFLQEFGLLPEKEVLVLTSVRNSATCHSEDSDLEIDSpecifications
AAH17227 | |
Liquid | |
5082 | |
Human PDCL Full-length ORF (AAH17227, 1 a.a. - 301 a.a.) Recombinant Protein with GST-tag at N-terminal | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at −80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp564M1863, PhLP | |
PDCL | |
Yes | |
wheat germ expression system |
AP, Array, ELISA, WB-Re | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.85kDa | |
Glutathione Sepharose 4 Fast Flow | |
25μg | |
RUO | |
PDCL | |
Wheat Germ (in vitro) | |
GST Tag |