missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PCDHB2 Full-length ORF (AAH98575.1, 1 a.a. - 798 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00056133-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132103

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. [provided by RefSeq]

Sequence: MEAGEGKERVPKQRQVLIFFVLLGIAQASCQPRHYSVAEETESGSFVANLLKDLGLEIGELAVRGARVVSKGKKMHLQFDRQTGDLLLNEKLDREELCGPTEPCVLPFQVLLENPLQFFQAELRIRDVNDHSPVFLDKEILLKIPESITPGTTFLIERAQDLDVGTNSLQNYTISPNFHFHLNLQDSLDGIILPQLVLNRALDREEQPEIRLTLTALDGGSPPRSGTALVRIEVVDINDNVPEFAKLLYEVQIPEDSPVGSQVAIVSARDLDIGTNGEISYAFSQASEDIRKTFRLSAKSGELLLRQKLDFESIQTYTVNIQATDGGGLSGTCVVFVQVMDLNDNPPELTMSTLINQIPENLQDTLIAVFSVSDPDSGDNGRMVCSIQDDLPFFLKPSVENFYTLVISTALDRETRSEYNITITVTDFGTPRLKTEHNITVLVSDVNDNAPAFTQTSYTLFVRENNSPALHIGSVSATDRDSGTNAQVTYSLLPPQDPHLPLASLVSINADNGHLFALQSLDYEALQAFEFRVGAADRGSPALSSEALVRVLVLDANDNSPFVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATEPGLFGVWAHNGEVRTARLLRERDAAKQRLVVLVKDNGEPPRSATATLHVLLVDGFSQPYLLLPEAAPAQAQADLLTVYLVVALASVSSLFLFSVLLFVAVRLCRRSRAASVGRCSVPEGPFPGQMVDVSGTGTLSQSYQYEVCLTGESGTNEFKFLKPIIPNFVAQGAERVSEANPSFRKSFEFT
Specifications

Specifications

AAH98575.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
113.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC111392/PCDH-BETA2
PCDHB2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
56133
PCDHB2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEAGEGKERVPKQRQVLIFFVLLGIAQASCQPRHYSVAEETESGSFVANLLKDLGLEIGELAVRGARVVSKGKKMHLQFDRQTGDLLLNEKLDREELCGPTEPCVLPFQVLLENPLQFFQAELRIRDVNDHSPVFLDKEILLKIPESITPGTTFLIERAQDLDVGTNSLQNYTISPNFHFHLNLQDSLDGIILPQLVLNRALDREEQPEIRLTLTALDGGSPPRSGTALVRIEVVDINDNVPEFAKLLYEVQIPEDSPVGSQVAIVSARDLDIGTNGEISYAFSQASEDIRKTFRLSAKSGELLLRQKLDFESIQTYTVNIQATDGGGLSGTCVVFVQVMDLNDNPPELTMSTLINQIPENLQDTLIAVFSVSDPDSGDNGRMVCSIQDDLPFFLKPSVENFYTLVISTALDRETRSEYNITITVTDFGTPRLKTEHNITVLVSDVNDNAPAFTQTSYTLFVRENNSPALHIGSVSATDRDSGTNAQVTYSLLPPQDPHLPLASLVSINADNGHLFALQSLDYEALQAFEFRVGAADRGSPALSSEALVRVLVLDANDNSPFVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATEPGLFGVWAHNGEVRTARLLRERDAAKQRLVVLVKDNGEPPRSATATLHVLLVDGFSQPYLLLPEAAPAQAQADLLTVYLVVALASVSSLFLFSVLLFVAVRLCRRSRAASVGRCSVPEGPFPGQMVDVSGTGTLSQSYQYEVCLTGESGTNEFKFLKPIIPNFVAQGAERVSEANPSFRKSFEFT
RUO
PCDHB2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers