missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PAFAH1B1 Full-length ORF (AAH64638.1, 1 a.a. - 410 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005048-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132325

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This locus was identified as encoding a gene that when mutated or lost caused the lissencephaly associated with Miller-Dieker lissencephaly syndrome. This gene encodes the non-catalytic alpha subunit of the intracellular Ib isoform of platelet-activating factor acteylhydrolase, a heterotrimeric enzyme that specifically catalyzes the removal of the acetyl group at the SN-2 position of platelet-activating factor (identified as 1-O-alkyl-2-acetyl-sn-glyceryl-3-phosphorylcholine). Two other isoforms of intracellular platelet-activating factor acetylhydrolase exist: one composed of multiple subunits, the other, a single subunit. In addition, a single-subunit isoform of this enzyme is found in serum. [provided by RefSeq]

Sequence: MVLSQRQRDELNRAIADYLRSNGYEEAYSVFKKEAELDVNEELDKKYAGLLEKKWTSVIRLQKKVMELESKLNEAKEEFTSGGPLGQKRDPKEWIPRPPEKYALSGHRSPVTRVIFHPVFSVMVSASEDATIKVWDYETGDFERTLKGHTDSVQDISFDHSGKLLASCSADMTIKLWDFQGFECIRTMHGHDHNVSSVAIMPNGDHIVSASRDKTIKMWEVQTGYCVKTFTGHREWVRMVRPNQDGTLIASCSNDQTVRVWVVATKECKAELREHEHVVECISWAPESSYSSISEATGSETKKSGKPGPFLLSGSRDKTIKMWDVSTGMCLMTLVGHDNWVRGVLFHSGGKFILSCADDKTLRVWDYKNKRCMKTLNAHEHFVTSLDFHKTAPYVVTGSVDQTVKVWECR
Specifications

Specifications

AAH64638.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LIS1/LIS2/MDCR/MDS/PAFAH
PAFAH1B1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5048
PAFAH1B1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVLSQRQRDELNRAIADYLRSNGYEEAYSVFKKEAELDVNEELDKKYAGLLEKKWTSVIRLQKKVMELESKLNEAKEEFTSGGPLGQKRDPKEWIPRPPEKYALSGHRSPVTRVIFHPVFSVMVSASEDATIKVWDYETGDFERTLKGHTDSVQDISFDHSGKLLASCSADMTIKLWDFQGFECIRTMHGHDHNVSSVAIMPNGDHIVSASRDKTIKMWEVQTGYCVKTFTGHREWVRMVRPNQDGTLIASCSNDQTVRVWVVATKECKAELREHEHVVECISWAPESSYSSISEATGSETKKSGKPGPFLLSGSRDKTIKMWDVSTGMCLMTLVGHDNWVRGVLFHSGGKFILSCADDKTLRVWDYKNKRCMKTLNAHEHFVTSLDFHKTAPYVVTGSVDQTVKVWECR
RUO
PAFAH1B1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers