Learn More
Abnova™ Human P2RX3 Partial ORF (NP_002550, 45 a.a. - 154 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005024-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel and may transduce ATP-evoked nociceptor activation. Mouse studies suggest that this receptor is important for peripheral pain responses, and also participates in pathways controlling urinary bladder volume reflexes. It is possible that the development of selective antagonists for this receptor may have a therapeutic potential in pain relief and in the treatment of disorders of urine storage. [provided by RefSeq]
Sequence: HEKAYQVRDTAIESSVVTKVKGSGLYANRVMDVSDYVTPPQGTSVFVIITKMIVTENQMQGFCPESEEKYRCVSDSQCGPERLPGGGILTGRCVNYSSVLRTCEIQGWCPSpecifications
NP_002550 | |
Liquid | |
5024 | |
P2RX3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC129956/P2X3 | |
P2RX3 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HEKAYQVRDTAIESSVVTKVKGSGLYANRVMDVSDYVTPPQGTSVFVIITKMIVTENQMQGFCPESEEKYRCVSDSQCGPERLPGGGILTGRCVNYSSVLRTCEIQGWCP | |
RUO | |
P2RX3 | |
Wheat Germ (in vitro) | |
GST |