missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OR4K2 Full-length ORF (NP_001005501.1, 1 a.a. - 314 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00390431-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140564

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]

Sequence: MDVGNKSTMSEFVLLGLSNSWELQMFFFMVFSLLYVATMVGNSLIVITVIVDPHLHSPMYFLLTNLSIIDMSLASFATPKMITDYLTGHKTISFDGCLTQIFFLHLFTGTEIILLMAMSFDRYIAICKPLHYASVISPQVCVALVVASWIMGVMHSMSQVIFALTLPFCGPYEVDSFFCDLPVVFQLACVDTYVLGLFMISTSGIIALSCFIVLFNSYVIVLVTVKHHSSRGSSKALSTCTAHFIVVFLFFGPCIFIYMWPLSSFLTDKILSVFYTIFTPTLNPIIYTLRNQEVKIAMRKLKNRFLNFNKAMPS
Specifications

Specifications

NP_001005501.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OR14-15
OR4K2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
390431
OR4K2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDVGNKSTMSEFVLLGLSNSWELQMFFFMVFSLLYVATMVGNSLIVITVIVDPHLHSPMYFLLTNLSIIDMSLASFATPKMITDYLTGHKTISFDGCLTQIFFLHLFTGTEIILLMAMSFDRYIAICKPLHYASVISPQVCVALVVASWIMGVMHSMSQVIFALTLPFCGPYEVDSFFCDLPVVFQLACVDTYVLGLFMISTSGIIALSCFIVLFNSYVIVLVTVKHHSSRGSSKALSTCTAHFIVVFLFFGPCIFIYMWPLSSFLTDKILSVFYTIFTPTLNPIIYTLRNQEVKIAMRKLKNRFLNFNKAMPS
RUO
OR4K2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers