Få mere at vide
Gibco™ Human NT-3 Recombinant Protein, PeproTech®
Artikelnummer. 17824623
Mængde:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Pakningsstørrelse:
50µg
Artikelnummer. 17824623
Brand: Gibco™ 450032UG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Recombinant Protein
Recombinant Human NT-3 is a noncovalently linked homodimer of two 13.6 kDa polypeptide monomers (240 total amino acid residues). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.
TRUSTED_SUSTAINABILITY
Tekniske data
Adgangsnummer P20783
Til brug med (applikation) ELISA, Functional Assay, Western Blot
Formulering protein with no preservative
Gen-id (Entrez) 4908
Molekylvægt (g/mol) 27.2 kDa
Navn Human NT-3
Mængde 2 μg
Kilde E. coli
Regulatorisk status RUO
Endotoksin koncentration <1 EU/ μg
Gene Alias AI316846; AI835689; HDNF; Nerve growth factor 2; neurotrophic factor; neurotrophin 3; neurotrophin-3; neurotrophin-3 (HDNF/NT-3); NGF2; NGF-2; NT3; NT-3; NTF3; Ntf-3
Fælles navn NTF3
Gen symbol Ntf3
Biologisk aktivitet Determined by its ability to bind recombinant human TrkB Fc Chimera in a functional ELISA.
Konjugeret Unconjugated
Rekombinant Recombinant
Sekvens MYAEHKSHRG EYSVCDSESL WVTDKSSAID IRGHQVTVLG EIKTGNSPVK QYFYETRCKE ARPVKNGCRG IDDKHWNSQC KTSQTYVRAL TSENNKLVGW RWIRIDTSCV CALSRKIGRT
Indhold og opbevaring -20°C
Udtrykssystem E. coli
Form Lyophilized
Renhed eller kvalitetsklasse ≥ 98% by SDS-PAGE gel and HPLC analyses.