missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NSMAF Partial ORF (NP_003571, 818 a.a. - 917 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_003571 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8439 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16148535
|
Abnova™
H00008439-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16138535
|
Abnova™
H00008439-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a WD-repeat protein that binds the cytoplasmic sphingomyelinase activation domain of the 55kD tumor necrosis factor receptor. This protein is required for TNF-mediated activation of neutral sphingomyelinase and may play a role in regulating TNF-induced cellular responses such as inflammation. Alternative splicing results in multiple transcript variants
Sequence: RHVLSTGTDGCLNVIDVQTGMLISSMTSDEPQRCFVWDGNSVLSGSQSGELLVWDLLGAKISERIQGHTGAVTCIWMNEQCSSIITGGEDRQIIFWKLQYSpecifications
NP_003571 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FAN | |
NSMAF | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8439 | |
NSMAF (Human) Recombinant Protein (Q01) | |
RHVLSTGTDGCLNVIDVQTGMLISSMTSDEPQRCFVWDGNSVLSGSQSGELLVWDLLGAKISERIQGHTGAVTCIWMNEQCSSIITGGEDRQIIFWKLQY | |
RUO | |
NSMAF | |
Wheat Germ (in vitro) | |
GST | |
Liquid |