missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NRXN3 Partial ORF (NP_620426, 101 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009369-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180486

  • 2665.00 DKK / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Neurexins are a family of proteins that function in the vertebrate nervous system as cell adhesion molecules and receptors. They are encoded by several unlinked genes of which two, NRXN1 and NRXN3, are among the largest known human genes. Three of the genes (NRXN1-3) utilize two alternate promoters and include numerous alternatively spliced exons to generate thousands of distinct mRNA transcripts and protein isoforms. The majority of transcripts are produced from the upstream promoter and encode alpha-neurexin isoforms; a much smaller number of transcripts are produced from the downstream promoter and encode beta-neurexin isoforms. The alpha-neurexins contain epidermal growth factor-like (EGF-like) sequences and laminin G domains, and have been shown to interact with neurexophilins. The beta-neurexins lack EGF-like sequences and contain fewer laminin G domains than alpha-neurexins. [provided by RefSeq]

Sequence: PANDRPSTRSDRLAVGFSTTVKDGILVRIDSAPGLGDFLQLHIEQGKIGVVFNIGTVDISIKEERTPVNDGKYHVVRFTRNGGNATLQVDNWPVNEHYPTGRQLTIFNTQ
Specifications

Specifications

NP_620426
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PANDRPSTRSDRLAVGFSTTVKDGILVRIDSAPGLGDFLQLHIEQGKIGVVFNIGTVDISIKEERTPVNDGKYHVVRFTRNGGNATLQVDNWPVNEHYPTGRQLTIFNTQ
RUO
NRXN3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9369
NRXN3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0743/MGC176711
NRXN3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers