missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NMI Partial ORF (AAH01268, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009111-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189945

  • 4040.00 DKK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NMYC interactor (NMI) encodes a protein that interacts with NMYC and CMYC (two members of the oncogene Myc family), and other transcription factors containing a Zip, HLH, or HLH-Zip motif. The NMI protein also interacts with all STATs except STAT2 and augments STAT-mediated transcription in response to cytokines IL2 and IFN-gamma. The NMI mRNA has low expression levels in all human fetal and adult tissues tested except brain and has high expression in cancer cell line-myeloid leukemias. [provided by RefSeq]

Sequence: MEADKDDTQQILKEHSPDEFIKDEQNKGLIDEITKKNIQLKKEIQKLETELQEATKEFQIKEDIPETKMKFLSVETPENDSQLSNISCSFQVSSKVPYEI
Specifications

Specifications

AAH01268
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MEADKDDTQQILKEHSPDEFIKDEQNKGLIDEITKKNIQLKKEIQKLETELQEATKEFQIKEDIPETKMKFLSVETPENDSQLSNISCSFQVSSKVPYEI
RUO
NMI
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
9111
NMI (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NMI
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers