Learn More
Abnova™ Human NLGN4X Partial ORF (NP_065793, 577 a.a. - 676 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057502-Q01.10ug
Description
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. The encoded protein interacts with discs, large (Drosophila) homolog 4 (DLG4). Mutations in this gene have been associated with autism and Asperger syndrome. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq]
Sequence: RVRDHYRATKVAFWLELVPHLHNLNEIFQYVSTTTKVPPPDMTSFPYGTRRSPAKIWPTTKRPAITPANNPKHSKDPHKTGPEDTTVLIETKRDYSTELSSpecifications
NP_065793 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RVRDHYRATKVAFWLELVPHLHNLNEIFQYVSTTTKVPPPDMTSFPYGTRRSPAKIWPTTKRPAITPANNPKHSKDPHKTGPEDTTVLIETKRDYSTELS | |
RUO | |
NLGN4X | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57502 | |
NLGN4X (Human) Recombinant Protein (Q01) | |
10 ÎĽg | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASPGX2/AUTSX2/HLNX/HNLX/KIAA1260/MGC22376/NLGN/NLGN4 | |
NLGN4X | |
Recombinant | |
wheat germ expression system |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.