missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Abnova™ Human NID2 Partial ORF (NP_031387.2, 1276 a.a. - 1375 a.a.) Recombinant Protein with GST-tag at N-terminal
Klik for at se tilgængelige muligheder
Mængde:
10 μg
25 μg
Pakningsstørrelse:
25µg
Beskrivelse
Basement membranes, which are composed of type IV collagens (see MIM 120130), laminins (see LAMC1; MIM 150290), perlecan (HSPG2; MIM 142461), and nidogen (see NID1; MIM 131390), are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis.[supplied by OMIM]
Sequence: PNGLTFDPFSKLLCWADAGTKKLECTLPDGTGRRVIQNNLKYPFSIVSYADHFYHTDWRRDGVVSVNKHSGQFTDEYLPEQRSHLYGITAVYPYCPTGRK
Tekniske data
Tekniske data
| Adgangsnummer | NP_031387.2 |
| Til brug med (applikation) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulering | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gen-id (Entrez) | 22795 |
| Molekylvægt (g/mol) | 36.74kDa |
| Navn | NID2 (Human) Recombinant Protein (Q01) |
| Kvalitetskontrol test | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Mængde | 25 μg |
| Immunogen | PNGLTFDPFSKLLCWADAGTKKLECTLPDGTGRRVIQNNLKYPFSIVSYADHFYHTDWRRDGVVSVNKHSGQFTDEYLPEQRSHLYGITAVYPYCPTGRK |
| Opbevaringskrav | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Abnova™ Human NID2 Partial ORF (NP_031387.2, 1276 a.a. - 1375 a.a.) Recombinant Protein with GST-tag at N-terminal >
Ser du en mulighed for forbedring?Del en indholdskorrektion