missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NIBP Full-length ORF (AAH06206.3, 1 a.a. - 912 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00083696-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135433

  • 4040.00 DKK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NIBP binds NIK (MIM 604655) and IKK-beta (IKBKB; MIM 603258) and plays a role in the neuronal NF-kappa-B (see MIM 164011) signaling pathway (Hu et al., 2005 [PubMed 15951441]).[supplied by OMIM]

Sequence: MSVELLRSVNDFLWLGAALEGLCSASVIYHYPGGTGGKSGARRFQGSTLPAEAANRHRPGALTTNGINPDTSTEIGRAKNCLSPEDIIDKYKEAISYYSKYKNAGVIELEACIKAVRVLAIQKRSMEASEFLQNAVYINLRQLSEEEKIQRYSILSELYELIGFHRKSAFFKRVAAMQCVAPSIAEPGWRACYKLLLETLPGYSLSLDPKDFSRGTHRGWAAVQMRLLHELVYASRRMGNPALSVRHLSFLLQTMLDFLSDQEKKDVAQSLENYTSKCPGTMEPIALPGGLTLPPVPFTKLPIVRHVKLLNLPASLRPHKMKSLLGQNVSTKSPFIYSPIIAHNRGEERNKKIDFQWVQGDVCEVQLMVYNPMPFELRVENMGLLTSGVEFESLPAALSLPAESGLYPVTLVGVPQTTGTITVNGYHTTVFGVFSDCLLDNLPGIKTSGSTVEVIPALPRLQISTSLPRSAHSLQPSSGDEISTNVSVQLYNGESQQLIIKLENIGMEPLEKLEVTSKVLTTKEKLYGDFLSWKLEETLAQFPLQPGKVATFTINIKVKLDFSCQENLLQDLSDDGISVSGFPLSSPFRQVVRPRVEGKPVNPPESNKAGDYSHVKTLEAVLNFKYSGGPGHTEGYYRNLSLGLHVEVEPSVFFTRVSTLPATSTRQCHLLLDVFNSTEHELTVSTRSSEALILHAGECQRMAIQVDKFNFESFPESPGEKGQFANPKQLEEERREARGLEIHSKLGICWRIPSLKRSGEASVEGLLNQLVLEHLQLAPLQWDVLVDGQPCDREAVAACQVGDPVRLEVRLTNRSPRSVGPFALTVVPFQDHQNGVHNYDLHDTVSFVGSSTFYLDAVQPSGQSACLGALLFLYTGDFFLHIRFHEDSTSKELPPSWFCLPSVHVCALEAQA
Specifications

Specifications

AAH06206.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
127.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IBP/IKBKBBP/KIAA1882/MGC4737/MGC4769/NIBP/T1/TRS120
TRAPPC9
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
83696
NIBP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSVELLRSVNDFLWLGAALEGLCSASVIYHYPGGTGGKSGARRFQGSTLPAEAANRHRPGALTTNGINPDTSTEIGRAKNCLSPEDIIDKYKEAISYYSKYKNAGVIELEACIKAVRVLAIQKRSMEASEFLQNAVYINLRQLSEEEKIQRYSILSELYELIGFHRKSAFFKRVAAMQCVAPSIAEPGWRACYKLLLETLPGYSLSLDPKDFSRGTHRGWAAVQMRLLHELVYASRRMGNPALSVRHLSFLLQTMLDFLSDQEKKDVAQSLENYTSKCPGTMEPIALPGGLTLPPVPFTKLPIVRHVKLLNLPASLRPHKMKSLLGQNVSTKSPFIYSPIIAHNRGEERNKKIDFQWVQGDVCEVQLMVYNPMPFELRVENMGLLTSGVEFESLPAALSLPAESGLYPVTLVGVPQTTGTITVNGYHTTVFGVFSDCLLDNLPGIKTSGSTVEVIPALPRLQISTSLPRSAHSLQPSSGDEISTNVSVQLYNGESQQLIIKLENIGMEPLEKLEVTSKVLTTKEKLYGDFLSWKLEETLAQFPLQPGKVATFTINIKVKLDFSCQENLLQDLSDDGISVSGFPLSSPFRQVVRPRVEGKPVNPPESNKAGDYSHVKTLEAVLNFKYSGGPGHTEGYYRNLSLGLHVEVEPSVFFTRVSTLPATSTRQCHLLLDVFNSTEHELTVSTRSSEALILHAGECQRMAIQVDKFNFESFPESPGEKGQFANPKQLEEERREARGLEIHSKLGICWRIPSLKRSGEASVEGLLNQLVLEHLQLAPLQWDVLVDGQPCDREAVAACQVGDPVRLEVRLTNRSPRSVGPFALTVVPFQDHQNGVHNYDLHDTVSFVGSSTFYLDAVQPSGQSACLGALLFLYTGDFFLHIRFHEDSTSKELPPSWFCLPSVHVCALEAQA
RUO
TRAPPC9
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers