Få mere at vide
Novus Biologicals™ Human Neurilemmoma Whole Tissue Lysate (Adult Whole Tumor)
Shop alle Bio Techne produkterBeskrivelse
The Human Neurilemmoma Whole Tissue Lysate (Adult Whole Tumor) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Tekniske data
Tekniske data
| Til brug med (applikation) | Western Blot |
| Væv | Nervous |
| Mængde | 1 mg |
| Opbevaringsbuffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Testspecificitet | Total Protein - Human Tumor Tissue: Neurilemmoma |
| Cellulær fraktion | Whole |
| Vævstilstand | Tumor |
| Krydsreaktivitet | Human |
| Værtsarter | Human |
| Koncentration | 5 mg/mL |
| Vis mere |
For Research Use Only