missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NDUFV2 Partial ORF (NP_066552, 150 a.a. - 249 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004729-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138261

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: EAIQKKLGIKVGETTPDKLFTLIEVECLGACVNAPMVQINDNYYEDLTAKDIEEIIDELKAGKIPKPGPRSGRFSCEPAGGLTSLTEPPKGPGFGVQAGL
Specifications

Specifications

NP_066552
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EAIQKKLGIKVGETTPDKLFTLIEVECLGACVNAPMVQINDNYYEDLTAKDIEEIIDELKAGKIPKPGPRSGRFSCEPAGGLTSLTEPPKGPGFGVQAGL
RUO
NDUFV2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
4729
NDUFV2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NDUFV2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers