Learn More
Abnova™ Human NDUFB5 Full-length ORF (NP_002483.1, 1 a.a. - 189 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004711-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. [provided by RefSeq]
Sequence: MAAMSLLRRVSVTAVAALSGRPLGTRLGFGGFLTRGFPKAAAPVRHSGDHGKRLFVIRPSRFYDRRFLKLLRFYIALTGIPVAIFITLVNVFIGQAELAEIPEGYVPEHWEYYKHPISRWIARNFYDSPEKIYERTMAVLQIEAEKAELRVKELEVRKLMHVRGDGPWYYYETIDKELIDHSPKATPDNSpecifications
NP_002483.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CI-SGDH/DKFZp686N02262/FLJ30597/MGC111204/MGC12314/SGDH | |
NDUFB5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4711 | |
NDUFB5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAMSLLRRVSVTAVAALSGRPLGTRLGFGGFLTRGFPKAAAPVRHSGDHGKRLFVIRPSRFYDRRFLKLLRFYIALTGIPVAIFITLVNVFIGQAELAEIPEGYVPEHWEYYKHPISRWIARNFYDSPEKIYERTMAVLQIEAEKAELRVKELEVRKLMHVRGDGPWYYYETIDKELIDHSPKATPDN | |
RUO | |
NDUFB5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |