missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MSL3L1 Partial ORF (NP_523353.1, 1 a.a. - 79 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_523353.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10943 |
Molecular Weight (g/mol) | 34.43kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16143416
|
Abnova™
H00010943-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16133416
|
Abnova™
H00010943-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a nuclear protein and has similarity to drosophila male-specific lethal-3 gene. The drosophila protein plays a critical role in a dosage-compensation pathway, which equalizes X-linked gene expression in males and females. Thus this encoded protein is thought to play a similar function in chromatin remodeling and transcriptional regulation. This gene has been found to undergo X inactivation. There are four alternatively spliced transcript variants of this gene encoding different isoforms. [provided by RefSeq]
Sequence: MSASEGMKFKFHSGEKVLCFEPDPTKARVLYDAKIVVVIVGKDEKGRKIPEYLIHFNGWNRSWDRWAAEDHVLRDTDENSpecifications
NP_523353.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.43kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586J1822/MSL3L1 | |
MSL3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10943 | |
MSL3L1 (Human) Recombinant Protein (Q01) | |
MSASEGMKFKFHSGEKVLCFEPDPTKARVLYDAKIVVVIVGKDEKGRKIPEYLIHFNGWNRSWDRWAAEDHVLRDTDEN | |
RUO | |
MSL3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |