missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MSC Partial ORF (NP_005089.1, 103 a.a. - 188 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_005089.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9242 |
Molecular Weight (g/mol) | 35.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16170246
|
Abnova™
H00009242-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16180246
|
Abnova™
H00009242-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a transcriptional repressor capable of binding an E-box element either as a homodimer or as a heterodimer with E2A in vitro. The encoded protein also forms heterodimers with E2A proteins in vivo. This protein is capable of inhibiting the transactivation capability of E47, an E2A protein, in mammalian cells. This gene is a downstream target of the B-cell receptor signal transduction pathway. [provided by RefSeq]
Sequence: ECKQSQRNAANARERARMRVLSKAFSRLKTSLPWVPPDTKLSKLDTLRLASSYIAHLRQLLQEDRYENGYVHPVNLTWPFVVSGRPSpecifications
NP_005089.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.2kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABF-1/ABF1/MYOR/bHLHa22 | |
MSC | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9242 | |
MSC (Human) Recombinant Protein (Q01) | |
ECKQSQRNAANARERARMRVLSKAFSRLKTSLPWVPPDTKLSKLDTLRLASSYIAHLRQLLQEDRYENGYVHPVNLTWPFVVSGRP | |
RUO | |
MSC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |