missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MPZL3 Full-length ORF (NP_938016.1, 1 a.a. - 235 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00196264-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139163

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MQQRGAAGSRGCALFPLLGVLFFQGVYIVFSLEIRADAHVRGYVGEKIKLKCTFKSTSDVTDKLTIDWTYRPPSSSHTVSIFHYQSFQYPTTAGTFRDRISWVGNVYKGDASISISNPTIKDNGTFSCAVKNPPDVHHNIPMTELTVTERGFGTMLSSVALLSILVFVPSAVVVALLLVRMGRKAAGLKKRSRSGYKKSSIEVSDDTDQEEEEACMARLCVRCAECLDSDYEETY
Specifications

Specifications

NP_938016.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
52.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MPZL3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
196264
MPZL3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQQRGAAGSRGCALFPLLGVLFFQGVYIVFSLEIRADAHVRGYVGEKIKLKCTFKSTSDVTDKLTIDWTYRPPSSSHTVSIFHYQSFQYPTTAGTFRDRISWVGNVYKGDASISISNPTIKDNGTFSCAVKNPPDVHHNIPMTELTVTERGFGTMLSSVALLSILVFVPSAVVVALLLVRMGRKAAGLKKRSRSGYKKSSIEVSDDTDQEEEEACMARLCVRCAECLDSDYEETY
RUO
MPZL3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers