Learn More
Abnova™ Human MPHOSPH10 Partial ORF (NP_005782, 348 a.a. - 457 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010199-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is phosphorylated during mitosis. The protein localizes to the nucleolus during interphase and to the chromosomes during M phase. The protein is thought to be part of the U3 small nucleolar ribonucleoprotein complex, which is involved in rRNA processing. [provided by RefSeq]
Sequence: NVKKNSDEVKSSFEKRQEKMNEKIASLEKELLEKKPWQLQGEVTAQKRPENSLLEETLHFDHAVRMAPVITEETTLQLEDIIKQRIRDQAWDDVVRKEKPKEDAYEYKKRSpecifications
NP_005782 | |
Liquid | |
10199 | |
MPHOSPH10 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MPP10/MPP10P | |
MPHOSPH10 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NVKKNSDEVKSSFEKRQEKMNEKIASLEKELLEKKPWQLQGEVTAQKRPENSLLEETLHFDHAVRMAPVITEETTLQLEDIIKQRIRDQAWDDVVRKEKPKEDAYEYKKR | |
RUO | |
MPHOSPH10 | |
Wheat Germ (in vitro) | |
GST |