missing translation for 'onlineSavingsMsg'
Få mere at vide

Gibco™ Human/Mouse/Rat BMP-2 Recombinant Protein, PeproTech®

Artikelnummer. 17830923
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 17830923

Brand: Gibco™ 1200210UG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Edge
Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

Recombinant Human/Murine/Rat BMP-2 is a 26.0 kDa homodimeric protein consisting of two 115 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Bone Morphogenic Proteins (BMP) are members of the TGF-beta superfamily that affect bone and cartilage formation (Hogan 1996, Reddi 1998 and Francis-West et al. 1999). Mature BMPs are 30-38 kDa proteins that assume a TGF-beta -like cysteine knot configuration. Lovostatin increases bone formation by turning on the bmp-2 gene (Mundy et al. 1999). BMPs stimulate the production of specific bone matrix proteins and alter stromal cell and osteoclast proliferation (Macias et al. 1999, Lecanda et al. 1997). BMPs may also be an important factor for development of the viscera, with roles in cell proliferation, apoptosis, differentiation, and morphogenesis (Hogan 1996, Dale and Wardle 1999). BMPs appear to be responsible for normal dorsal/ventral patterning. Like TGF-beta, BMPs bind to a type II receptor, which then recruits the transducing type I receptor unit, activating the Smad protein signaling pathway (Massague 1994, Derynck 1997, Attisano 1993).
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer P12643, P21274, P49001
Til brug med (applikation) ELISA, Functional Assay, Western Blot
Formulering protein with no preservative
Gen-id (Entrez) 12156, 29373, 650
Molekylvægt (g/mol) 26 kDa
Navn Human/Mouse/Rat BMP-2
Mængde 10 μg
Kilde E. coli
Regulatorisk status RUO
Endotoksin koncentration <1 EU/ μg
Gene Alias 2610024H22Rik; AI467020; AL117858; AW546137; BB189135; BDA2; BMP; BMP2; BMP-2; BMP2A; BMP-2 A; BMPRII; BMPR-II; Bone morphogenetic protein; Bone morphogenetic protein 2; bone morphogenetic protein 2 precursor (BMP-2) (BMP-2 A); bone morphogenetic protein 2 A; H-BMP-2; morphogen
Fælles navn BMP-2
Gen symbol BMP2
Biologisk aktivitet Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 0.5-1.0 ug/ml
Konjugeret Unconjugated
Rekombinant Recombinant
Sekvens MQAKHKQRKR LKSSCKRHPL YVDFSDVGWN DWIVAPPGYH AFYCHGECPF PLADHLNSTN HAIVQTLVNS VNSKIPKACC VPTELSAISM LYLDENEKVV LKNYQDMVVE GCGCR
Indhold og opbevaring -20°C
Udtrykssystem E. coli
Form Lyophilized
Renhed eller kvalitetsklasse ≥ 98% by SDS-PAGE gel and HPLC analyses.
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.