missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human/Mouse Lipoprotein Lipase/LPL Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF7197-SP
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Lipoprotein Lipase/LPL Polyclonal specifically detects Lipoprotein Lipase/LPL in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Tekniske data
| Lipoprotein Lipase/LPL | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.1, HDLCQ11, LIPD, LIPDEC 3.1.1.34, lipoprotein lipase, LPL | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4023 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P06858 | |
| LPL | |
| E. coli-derived recombinant human Lipoprotein Lipase/LPL Accession # P06858 | |
| 25 μg | |
| Primary | |
| Detects human and mouse Lipoprotein Lipase/LPL in Western blots. Detects human Lipoprotein Lipase/LPL in direct ELISAs and less than 1% cross-reactivity with recombinant human (rh) LIPG, rhLIPI, and rhPNLIPRP1 is observed. | |
| Human, Mouse | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion