missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MOGAT3 Full-length ORF (NP_835470.1, 1 a.a. - 341 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_835470.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 346606 |
Molecular Weight (g/mol) | 65.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180274
|
Abnova™
H00346606-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16190274
|
Abnova™
H00346606-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Acyl-CoA:monoacylglycerol acyltransferase (MOGAT; EC 2.3.1.22) catalyzes the synthesis of diacylglycerol from 2-monoacylglycerol and fatty acyl-CoA (Cheng et al., 2003 [PubMed 12618427]).[supplied by OMIM]
Sequence: MGVATTLQPPTTSKTLQKQHLEAVGAYQYVLTFLFMGPFFSLLVFVLLFTSLWPFSVFYLVWLYVDWDTPNQGGRRSEWIRNRAIWRQLRDYYPVKLVKTAELPPDRNYVLGAHPHGIMCTGFLCNFSTESNGFSQLFPGLRPWLAVLAGLFYLPVYRDYIMSFGLCPVSRQSLDFILSQPQLGQAVVIMVGGAHEALYSVPGEHCLTLQKRKGFVRLALRHGASLVPVYSFGENDIFRLKAFATGSWQHWCQLTFKKLMGFSPCIFWGRGLFSATSWGLLPFAVPITTVVGRPIPVPQRLHPTEEEVNHYHALYMTALEQLFEEHKESCGVPASTCLTFISpecifications
NP_835470.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGVATTLQPPTTSKTLQKQHLEAVGAYQYVLTFLFMGPFFSLLVFVLLFTSLWPFSVFYLVWLYVDWDTPNQGGRRSEWIRNRAIWRQLRDYYPVKLVKTAELPPDRNYVLGAHPHGIMCTGFLCNFSTESNGFSQLFPGLRPWLAVLAGLFYLPVYRDYIMSFGLCPVSRQSLDFILSQPQLGQAVVIMVGGAHEALYSVPGEHCLTLQKRKGFVRLALRHGASLVPVYSFGENDIFRLKAFATGSWQHWCQLTFKKLMGFSPCIFWGRGLFSATSWGLLPFAVPITTVVGRPIPVPQRLHPTEEEVNHYHALYMTALEQLFEEHKESCGVPASTCLTFI | |
RUO | |
MOGAT3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
346606 | |
MOGAT3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DC7/DGAT2L7/MGAT3/MGC119203/MGC119204 | |
MOGAT3 | |
Recombinant | |
wheat germ expression system |