Learn More
Abnova™ Human MNAT1 Full-length ORF (NP_002422.1, 1 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004331-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Cyclin-dependent kinases (CDKs), which play an essential role in cell cycle control of eukaryotic cells, are phosphorylated and thus activated by the CDK-activating kinase (CAK). CAK is a multisubunit protein that includes CDK7 (MIM 601955), cyclin H (CCNH; MIM 601953), and MAT1. MAT1 (for 'menage a trois-1') is involved in the assembly of the CAK complex.[supplied by OMIM]
Sequence: MDDQGCPRCKTTKYRNPSLKLMVNVCGHTLCESCVDLLFVRGAGNCPECGTPLRKSNFRVQLFEDPTVDKEVEIRKKVLKIYNKREEDFPSLREYNDFLEEVEEIVFNLTNNVDLDNTKKKMEIYQKENKDVIQKNKLKLTREQEELEEALEVERQENEQRRLFIQKEEQLQQILKRKNKQAFLDELESSDLPVALLLAQHKDRSTQLEMQLEKPKPVKPVTFSTGIKMGQHISLAPIHKLEEALYEYQPLQIETYGPHVPELEMLGRLGYLNHVRAASPQDLAGGYTSSLACHRALQDAFSGLFWQPSSpecifications
NP_002422.1 | |
Liquid | |
4331 | |
MNAT1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDDQGCPRCKTTKYRNPSLKLMVNVCGHTLCESCVDLLFVRGAGNCPECGTPLRKSNFRVQLFEDPTVDKEVEIRKKVLKIYNKREEDFPSLREYNDFLEEVEEIVFNLTNNVDLDNTKKKMEIYQKENKDVIQKNKLKLTREQEELEEALEVERQENEQRRLFIQKEEQLQQILKRKNKQAFLDELESSDLPVALLLAQHKDRSTQLEMQLEKPKPVKPVTFSTGIKMGQHISLAPIHKLEEALYEYQPLQIETYGPHVPELEMLGRLGYLNHVRAASPQDLAGGYTSSLACHRALQDAFSGLFWQPS | |
RUO | |
MNAT1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAT1/RNF66/TFB3 | |
MNAT1 | |
Yes | |
wheat germ expression system |