missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MMAB Full-length ORF (NP_443077.1, 1 a.a. - 250 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00326625-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190094

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein that catalyzes the final step in the conversion of vitamin B(12) into adenosylcobalamin (AdoCbl), a vitamin B12-containing coenzyme for methylmalonyl-CoA mutase. Mutations in the gene are the cause of vitamin B12-dependent methylmalonic aciduria linked to the cblB complementation group. [provided by RefSeq]

Sequence: MAVCGLGSRLGLGSRLGLRGCFGAARLLYPRFQSRGPQGVEDGDRPQPSSKTPRIPKIYTKTGDKGFSSTFTGERRPKDDQVFEAVGTTDELSSAIGFALELVTEKGHTFAEELQKIQCTLQDVGSALATPCSSAREAHLKYTTFKAGPILELEQWIDKYTSQLPPLTAFILPSGGKISSALHFCRAVCRRAERRVVPLVQMGETDANVAKFLNRLSDYLFTLARYAAMKEGNQEKIYMKNDPSAESEGL
Specifications

Specifications

NP_443077.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
53.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATR/MGC20496
MMAB
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
326625
MMAB (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAVCGLGSRLGLGSRLGLRGCFGAARLLYPRFQSRGPQGVEDGDRPQPSSKTPRIPKIYTKTGDKGFSSTFTGERRPKDDQVFEAVGTTDELSSAIGFALELVTEKGHTFAEELQKIQCTLQDVGSALATPCSSAREAHLKYTTFKAGPILELEQWIDKYTSQLPPLTAFILPSGGKISSALHFCRAVCRRAERRVVPLVQMGETDANVAKFLNRLSDYLFTLARYAAMKEGNQEKIYMKNDPSAESEGL
RUO
MMAB
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers