missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MGC42638 Full-length ORF (AAH40290.1, 1 a.a. - 75 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

2665.00 DKK - 4040.00 DKK

Specifications

Accession Number AAH40290.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 340561
Molecular Weight (g/mol) 35.2kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16180204
View Documents Promotion Details
Abnova™
H00340561-P01.25UG
25 ug
4040.00 DKK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16170204
View Documents Promotion Details
Abnova™
H00340561-P01.10UG
10 ug
2665.00 DKK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence: MALKLIHKEFLELARDPQPHCSAGPVWDDMLHWQATITRPNDSSYLGGVFFLKFPSDYLFKPPKIKFTNGIYHQR
Specifications

Specifications

AAH40290.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.2kDa
Glutathione Sepharose 4 Fast Flow
MALKLIHKEFLELARDPQPHCSAGPVWDDMLHWQATITRPNDSSYLGGVFFLKFPSDYLFKPPKIKFTNGIYHQR
RUO
UBE2DNL
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
340561
MGC42638 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC42638
UBE2DNL
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications