missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MFRP Partial ORF (NP_113621, 480 a.a. - 579 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_113621 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 83552 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16140487
|
Abnova™
H00083552-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16130487
|
Abnova™
H00083552-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the frizzled-related proteins. The encoded protein may play a role in eye development, as mutations in this gene have been associated with nanophthalmos, posterior microphthalmia, retinitis pigmentosa, foveoschisis, and optic disc drusen. The protein is encoded by a bicistronic mRNA, which also encodes C1q and tumor necrosis factor related protein 5
Sequence: NTTAFPNIWVGMITQEEVVEVLSGYKSLTSLPCYQHFRRLLCGLLVPRCTPLGSVLPPCRSVCQEAEHQCQSGLALLGTPWPFNCNRLPEAADLEACAQPSpecifications
NP_113621 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C1QTNF5/FLJ30570/NNO2/rd6 | |
MFRP | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
83552 | |
MFRP (Human) Recombinant Protein (Q01) | |
NTTAFPNIWVGMITQEEVVEVLSGYKSLTSLPCYQHFRRLLCGLLVPRCTPLGSVLPPCRSVCQEAEHQCQSGLALLGTPWPFNCNRLPEAADLEACAQP | |
RUO | |
MFRP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |