missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MEI1 Full-length ORF (AAH32248.1, 1 a.a. - 238 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00150365-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138613

  • 4040.00 DKK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MLALTLAKADSPRTALLCSAWLLTASFSAQQHKGSLQVHQTLSVEMDQVLKALSFPKKKAALLSAAILCFLRTALRQSFSSALVALVPSGAQPLPATKDTVLAPLRMSQVRSLVIGLQNLLVQKDPLLSQACVGCLEALLDYLDARSPDIALHVASQPWNRFLLFTLLDAGENSFLRPEILRLMTLLQSMGHLADHSMAQTLQASLEGLPPSTSSGQPPLQDMLCLGGVAVSLSHIRN
Specifications

Specifications

AAH32248.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
52kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC40042
MEI1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
150365
MEI1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLALTLAKADSPRTALLCSAWLLTASFSAQQHKGSLQVHQTLSVEMDQVLKALSFPKKKAALLSAAILCFLRTALRQSFSSALVALVPSGAQPLPATKDTVLAPLRMSQVRSLVIGLQNLLVQKDPLLSQACVGCLEALLDYLDARSPDIALHVASQPWNRFLLFTLLDAGENSFLRPEILRLMTLLQSMGHLADHSMAQTLQASLEGLPPSTSSGQPPLQDMLCLGGVAVSLSHIRN
RUO
MEI1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers