missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MCM3AP Partial ORF (AAH13285, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008888-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179565

  • 4040.00 DKK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The minichromosome maintenance protein 3 (MCM3) is one of the MCM proteins essential for the initiation of DNA replication. The protein encoded by this gene is a MCM3 binding protein. It was reported to have phosphorylation-dependent DNA-primase activity, which was up-regulated in antigen immunization induced germinal center. This protein was demonstrated to be an acetyltransferase that acetylates MCM3 and plays a role in DNA replication. The mutagenesis of a nuclear localization signal of MCM3 affects the binding of this protein with MCM3, suggesting that this protein may also facilitate MCM3 nuclear localization. [provided by RefSeq]

Sequence: VRVARCCEDVCAHLVDLFLVEEIFQTAKETLQELQCFCKYLQRWREAVTARKKLRRQMRAFPAAPCCVDVSDRLRALAPSAECPIAEENLARGLLDLGHA
Specifications

Specifications

AAH13285
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VRVARCCEDVCAHLVDLFLVEEIFQTAKETLQELQCFCKYLQRWREAVTARKKLRRQMRAFPAAPCCVDVSDRLRALAPSAECPIAEENLARGLLDLGHA
RUO
MCM3AP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8888
MCM3AP (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ44336/FLJ45306/GANP/KIAA0572/MAP80
MCM3AP
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers