Learn More
Abnova™ Human MCM3AP Partial ORF (AAH13285, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008888-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The minichromosome maintenance protein 3 (MCM3) is one of the MCM proteins essential for the initiation of DNA replication. The protein encoded by this gene is a MCM3 binding protein. It was reported to have phosphorylation-dependent DNA-primase activity, which was up-regulated in antigen immunization induced germinal center. This protein was demonstrated to be an acetyltransferase that acetylates MCM3 and plays a role in DNA replication. The mutagenesis of a nuclear localization signal of MCM3 affects the binding of this protein with MCM3, suggesting that this protein may also facilitate MCM3 nuclear localization. [provided by RefSeq]
Sequence: VRVARCCEDVCAHLVDLFLVEEIFQTAKETLQELQCFCKYLQRWREAVTARKKLRRQMRAFPAAPCCVDVSDRLRALAPSAECPIAEENLARGLLDLGHASpecifications
AAH13285 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VRVARCCEDVCAHLVDLFLVEEIFQTAKETLQELQCFCKYLQRWREAVTARKKLRRQMRAFPAAPCCVDVSDRLRALAPSAECPIAEENLARGLLDLGHA | |
RUO | |
MCM3AP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8888 | |
MCM3AP (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ44336/FLJ45306/GANP/KIAA0572/MAP80 | |
MCM3AP | |
Recombinant | |
wheat germ expression system |