missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MASK Partial ORF (NP_057626, 331 a.a. - 415 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051765-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16055525

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The product of this gene is a member of the GCK group III family of kinases, which are a subset of the Ste20-like kinases. The encoded protein contains an amino-terminal kinase domain, and a carboxy-terminal regulatory domain that mediates homodimerization. The protein kinase localizes to the Golgi apparatus and is specifically activated by binding to the Golgi matrix protein GM130. It is also cleaved by caspase-3 in vitro, and may function in the apoptotic pathway. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]

Sequence: KKPDPKKVQNGAEQDLVQTLSCLSMIITPAFAELKQQDENNASRNQAIEELEKSIAVAEAACPGITDKMVKKLIEKFQKCSADES
Specifications

Specifications

NP_057626
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.09kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KKPDPKKVQNGAEQDLVQTLSCLSMIITPAFAELKQQDENNASRNQAIEELEKSIAVAEAACPGITDKMVKKLIEKFQKCSADES
RUO
RP6-213H19.1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
51765
MASK (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MASK/MST4
RP6-213H19.1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers