Learn More
Abnova™ Human MAN1A1 Partial ORF (NP_005898.2, 110 a.a. - 199 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004121-Q01.25ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a class I mammalian Golgi 1,2-mannosidase which is a type II transmembrane protein. This protein catalyzes the removal of 3 distinct mannose residues from peptide-bound Man(9)-GlcNAc(2) oligosaccharides and belongs to family 47 of glycosyl hydrolases. [provided by RefSeq]
Sequence: PGDPEAALEDNLARIRENHERALREAKETLQKLPEEIQRDILLEKKKVAQDQLRDKAPFRGLPPVDFVPPIGVESREPADAAIREKRAKISpecifications
NP_005898.2 | |
Liquid | |
4121 | |
MAN1A1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HUMM3/HUMM9/MAN9 | |
MAN1A1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PGDPEAALEDNLARIRENHERALREAKETLQKLPEEIQRDILLEKKKVAQDQLRDKAPFRGLPPVDFVPPIGVESREPADAAIREKRAKI | |
RUO | |
MAN1A1 | |
Wheat Germ (in vitro) | |
GST |