missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAGEC1 Partial ORF (NP_005453.2, 1 a.a. - 139 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_005453.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9947 |
Molecular Weight (g/mol) | 40.92kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16055185
|
Abnova™
H00009947-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16065185
|
Abnova™
H00009947-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the melanoma antigen gene (MAGE) family. The proteins of this family are tumor-specific antigens that can be recognized by autologous cytolytic T lymphocytes. This protein contains a large number of unique short repetitive sequences in front of the MAGE-homologous sequence, and therefore is about 800 aa longer than the other MAGE proteins. [provided by RefSeq]
Sequence: MGDKDMPTAGMPSLLQSSSESPQSCPEGEDSQSPLQIPQSSPESDDTLYPLQSPQSRSEGEDSSDPLQRPPEGKDSQSPLQIPQSSPEGDDTQSPLQNSQSSPEGKDSLSPLEISQSPPEGEDVQSPLQNPASSFFSSASpecifications
NP_005453.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.92kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CT7/MAGE-C1/MGC39366 | |
MAGEC1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9947 | |
MAGEC1 (Human) Recombinant Protein (Q01) | |
MGDKDMPTAGMPSLLQSSSESPQSCPEGEDSQSPLQIPQSSPESDDTLYPLQSPQSRSEGEDSSDPLQRPPEGKDSQSPLQIPQSSPEGDDTQSPLQNSQSSPEGKDSLSPLEISQSPPEGEDVQSPLQNPASSFFSSA | |
RUO | |
MAGEC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |