missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAGEA10 Full-length ORF (NP_001011543.1, 1 a.a. - 369 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004109-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179977

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: MPRAPKRQRCMPEEDLQSQSETQGLEGAQAPLAVEEDASSSTSTSSSFPSSFPSSSSSSSSSCYPLIPSTPEEVSADDETPNPPQSAQIACSSPSVVASLPLDQSDEGSSSQKEESPSTLQVLPDSESLPRSEIDEKVTDLVQFLLFKYQMKEPITKAEILESVIKNYEDHFPLLFSEASECMLLVFGIDVKEVDPTGHSFVLVTSLGLTYDGMLSDVQSMPKTGILILILSIIFIEGYCTPEEVIWEALNMMGLYDGMEHLIYGEPRKLLTQDWVQENYLEYRQVPGSDPARYEFLWGPRAHAEIRKMSLLKFLAKVNGSDPRSFPLWYEEALKDEEERAQDRIATTDDTTAMASASSSATGSFSYPE
Specifications

Specifications

NP_001011543.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
67.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MAGE10/MGC10599
MAGEA10
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
4109
MAGEA10 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPRAPKRQRCMPEEDLQSQSETQGLEGAQAPLAVEEDASSSTSTSSSFPSSFPSSSSSSSSSCYPLIPSTPEEVSADDETPNPPQSAQIACSSPSVVASLPLDQSDEGSSSQKEESPSTLQVLPDSESLPRSEIDEKVTDLVQFLLFKYQMKEPITKAEILESVIKNYEDHFPLLFSEASECMLLVFGIDVKEVDPTGHSFVLVTSLGLTYDGMLSDVQSMPKTGILILILSIIFIEGYCTPEEVIWEALNMMGLYDGMEHLIYGEPRKLLTQDWVQENYLEYRQVPGSDPARYEFLWGPRAHAEIRKMSLLKFLAKVNGSDPRSFPLWYEEALKDEEERAQDRIATTDDTTAMASASSSATGSFSYPE
RUO
MAGEA10
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers