Få mere at vide
R&D Systems™ Human Lysyl Oxidase Homolog 2 DuoSet ELISA
Artikelnummer. 30002095
Mængde:
5 Plates
Artikelnummer. 30002095
Brand: R&D Systems™ DY263905
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Indeholder
Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
Human Lysyl Oxidase Homolog 2 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human Lysyl Oxidase Homolog 2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Lysyl Oxidase Homolog 2/LOXL2.
TRUSTED_SUSTAINABILITY
Tekniske data
Analyseområde 0.375 to 24 ng/mL
Detekterbare analytter Lysyl Oxidase Homolog 2
Assay Solid Phase ELISA
Konjugeret Biotin
Krydsreaktivitet <50%cross-species reactivity observed with species tested
Immunoassay-kitformat 96-well Strip Plate
Produkttype ELISA Kit
Prøvetype Cell Culture Supernatant
Tilstrækkelig til To run ELISAs on at least 5 x 96-well plates
Målarter Human
Til brug med (applikation) ELISA
Gen-id (Entrez) 4017
Værtsarter Human
Mængde 5 Plates
Prøvevolumen 100 μL
Opbevaringskrav Store the unopened product at 2° to 8°C. Do not use past expiration date.