Få mere at vide
R&D Systems™ Human LTA4H (NP_000886.1) VersaClone cDNA
Tekniske data
Tekniske data
| Gene | Leukotriene A4 Hydrolase/LTA4H |
| Adgangsnummer | NP_000886.1 |
| Koncentration | 10 μg at 0.2 μg/μL |
| Formulering | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Indsæt type | 1849 bp |
| Gen-id (Entrez) | 4048 |
| Gene Alias | EC 3.3.2.6, Leukotriene A(4) hydrolase, leukotriene A4 hydrolase, leukotriene A-4 hydrolase, LTA4, LTA-4 hydrolase, LTA4H |
| Indhold og opbevaring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Arter | Human |
| Produktlinje | R&D Systems |
| Vis mere |