Learn More
Abnova™ Human LMO1 Partial ORF (NP_002306.1, 15 a.a. - 156 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004004-Q02.25ug
Additional Details : Weight : 0.02000kg
Description
LMO1 enocdes a cysteine-rich, two LIM domain transcriptional regulator. It is mapped to an area of consistent chromosomal translocation in chromosome 11, disrupting it in T-cell leukemia, although more rarely than the related gene, LMO2 is disrupted. [provided by RefSeq]
Sequence: VQPKGKQKGCAGCNRKIKDRYLLKALDKYWHEDCLKCACCDCRLGEVGSTLYTKANLILCRRDYLRLFGTTGNCAACSKLIPAFEMVMRARDNVYHLDCFACQLCNQRFCVGDKFFLKNNMILCQMDYEEGQLNGTFESQVQSpecifications
NP_002306.1 | |
Liquid | |
4004 | |
LMO1 (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC116692/RBTN1/RHOM1/TTG1 | |
LMO1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
41.25kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
VQPKGKQKGCAGCNRKIKDRYLLKALDKYWHEDCLKCACCDCRLGEVGSTLYTKANLILCRRDYLRLFGTTGNCAACSKLIPAFEMVMRARDNVYHLDCFACQLCNQRFCVGDKFFLKNNMILCQMDYEEGQLNGTFESQVQ | |
RUO | |
LMO1 | |
Wheat Germ (in vitro) | |
GST |