Learn More
Abcam Human LMNA (Lamin A) knockout HeLa cell line
Product Code. 32155176 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32155176 1,000,000 Cells Each
32154426 2 x 1,000,000 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32155176 Supplier Abcam Supplier No. ab2617871000000Cells_
 Restricted Item
Forventet forsendelse: 07-12-2026
LMNA KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 19 bp deletion in exon 4 and 1 bp deletion in exon 4 and 22 bp deletion in exon 4. To order both knockout and wild-type control cells: select 2 ...
LMNA KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 19 bp deletion in exon 4 and 1 bp deletion in exon 4 and 22 bp deletion in exon 4. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1,000,000 Cells
Regulatory Status RUO