missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LHX2 Partial ORF (NP_004780, 1 a.a. - 76 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

2665.00 DKK - 4040.00 DKK

Specifications

Accession Number NP_004780
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9355
Molecular Weight (g/mol) 34.1kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16130456
View Documents Promotion Details
Abnova™
H00009355-Q01.10UG
10 ug
2665.00 DKK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16140456
View Documents Promotion Details
Abnova™
H00009355-Q01.25UG
25 ug
4040.00 DKK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution. [provided by RefSeq]

Sequence: MLFHSLSGPEVHGVIDEMDRRAKSEAPAISSAIDRGDTETTMPSISSDRAALCAGCGGKISDRYYLLAVDKQWHMR
Specifications

Specifications

NP_004780
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.1kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LH2/MGC138390/hLhx2
LHX2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
9355
LHX2 (Human) Recombinant Protein (Q01)
MLFHSLSGPEVHGVIDEMDRRAKSEAPAISSAIDRGDTETTMPSISSDRAALCAGCGGKISDRYYLLAVDKQWHMR
RUO
LHX2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications