missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Human Latrophilin 3/LPHN3 (NP_001309175) VersaClone cDNA

Artikelnummer. 18730599
Skift visning
Klik for at se tilgængelige muligheder
Koncentration:
10 μg at 0.2 μg/μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Koncentration
18730599 10 μg at 0.2 μg/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18730599 Leverandør R&D Systems™ Leverandørnr. RDC2159
  • ONLINE EXCLUSIVE PRICE
    9345.00 DKK
    8838.90 DKK / 10µg
    Spare 506.10 DKK
    5% Slukket
 Begrænset vare
Forventet forsendelse: 14-10-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

R&D Systems™ Human Latrophilin 3/LPHN3 (NP_001309175) VersaClone cDNA is a member of the latrophilin subfamily of G-protein coupled receptors (GPCR).

The Human Latrophilin 3/LPHN3 (NP_001309175) VersaClone cDNA clone contains the complete ORF for Latrophilin 3/LPHN3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Latrophilin 3/LPHN3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Tekniske data

Gene hADGRL3
Adgangsnummer NP_001309175
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10 mM Tris-Cl
Indsæt type 3736bp
pH 8.5
Gen-id (Entrez) 23284
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Arter Human
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.