Få mere at vide
Human L1CAM Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems FAB7773S-100UG
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Flow Cytometry
Tekniske data
| L1CAM | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| antigen identified by monoclonal R1, CAML1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
| Mouse myeloma cell line NS0 derived Human L1CAM, (Ile20Glu1120) & (Arg864Glu1120), Accession # CAA42508 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2702C | |
| Flow Cytometry | |
| CAA42508 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3897 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG |