missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR2DS1/CD158h PE-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody has been used in 1 publication
1845.00 DKK - 3290.00 DKK
Specifications
Antigen | KIR2DS1/CD158h |
---|---|
Clone | 1127B |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18175251
|
R&D Systems
FAB8887P-100 |
100 Tests |
3290.00 DKK
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18732018
![]() |
R&D Systems
FAB8887P-025 |
25 Tests |
1845.00 DKK
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIR2DS1/CD158h Monoclonal specifically detects KIR2DS1/CD158h in Human samples. It is validated for Flow Cytometry.Specifications
KIR2DS1/CD158h | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
killer cell immunoglobulin-like receptor 2DS1, CD158a, CD158H, EB6ActI, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 1, p50.1 | |
KIR2DS1 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
1127B | |
Flow Cytometry | |
R-PE | |
Rabbit | |
Human | |
Q14954 | |
3806 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Detects human KIR2DS1/CD158h in flow cytometry. Clone 1127B recognizes KIR2DS1 and some alleles of KIR2DL1, both members of the killer cell immunoglobulin-like receptor (KIR) family. Because Clone 1127B displays partial cross-reactivity with KIR2DL1, co-staining with Mouse Anti-Human KIR2DL1 Clone 143211 (Catalog # FAB1844A) is recommended. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human KIR2DS1/CD158h PE-conjugated Antibody, R&D Systems™